missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNPO2L Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82021R
This item is not returnable.
View return policy
Description
SYNPO2L Polyclonal antibody specifically detects SYNPO2L in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| SYNPO2L | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 550 | |
| Synaptopodin 2-Like, Synaptopodin 2-Like Protein | |
| SYNPO2L antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human SYNPO2L. The immunogen is located within amino acids 140 - 190 of SYNPO2L. Amino Acid Squence: PATQEKPRRPRRRGPTRPT | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 79933 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction