missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYNRG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570724
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
May play a role in endocytosis and/or membrane trafficking at the trans-Golgi network (TGN). May act by linking the adapter protein complex AP-1 to other proteins.
Specifications
| SYNRG | |
| Polyclonal | |
| Unconjugated | |
| SYNRG | |
| adaptor-related protein complex 1 gamma subunit-binding protein 1; AP1 gamma subunit binding protein 1; AP1 subunit gamma-binding protein 1; Ap1gbp1; fj67h08; Gamma-synergin; MGC104959; synergin gamma; synergin, gamma; Syng; Synrg; wu:fj67h08 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11276 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UMZ2 | |
| SYNRG | |
| synthetic peptide directed towards the N-terminal region of human SYNRG. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction