missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYNRG Polyclonal Antibody
GREENER_CHOICE

Product Code. 15722751 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15722751 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15722751 Supplier Invitrogen™ Supplier No. PA554820

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QQQGFPMVSV MQPNMQGIMG MNYSSQMSQG PIAMQAGIPM GPMPAAGMPY LGQAPFLGMR PPGPQYTPDM QKQFAEEQQK RFEQQQKLLE EERKRRQFEE QKQKLRLLSS VKP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

May play a role in endocytosis and/or membrane trafficking at the trans-Golgi network (TGN). May act by linking the adapter protein complex AP-1 to other proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYNRG
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYNRG
Gene Accession No. Q9UMZ2
Gene Alias adaptor-related protein complex 1 gamma subunit-binding protein 1; AP1 gamma subunit binding protein 1; AP1 subunit gamma-binding protein 1; Ap1gbp1; fj67h08; Gamma-synergin; MGC104959; synergin gamma; synergin, gamma; Syng; Synrg; wu:fj67h08
Gene Symbols SYNRG
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SYNRG. Recombinant protein control fragment (Product #RP-93211).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11276
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.