missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntabulin/GOLSYN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 624.00
Specifications
| Antigen | Syntabulin/GOLSYN |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18423012
|
Novus Biologicals
NBP2-30921-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18166384
|
Novus Biologicals
NBP2-30921 |
0.1 mL |
€ 624.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Syntabulin/GOLSYN Polyclonal specifically detects Syntabulin/GOLSYN in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Syntabulin/GOLSYN | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Golgi-Localized Protein, Golgi-Localized Syntaphilin-Related Protein, GOLSYN A Protein, GOLSYN B Protein, GOLSYN C Protein, Implicated In Syntaxin Trafficking In Neurons, KIAA1472, Microtubule-Associated Protein, OCSYN, SNPHL, SYBU, syntabulin, Syntabulin (Syntaxin-Interacting), syntaphilin-like, Syntaxin-1-Binding Protein | |
| SYBU | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q9NX95 | |
| 55638 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: STPGANVLELLPIVMGQEEGSVVVERAVQTDVVPYSPAISELIQSVLQKLQDPCPSSLASPDESEPDSMESFPESLSALVVDLTPRNPNSAI | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title