missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaphilin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 391.65 - € 590.10
Specifications
| Antigen | Syntaphilin |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18462102
|
Novus Biologicals
NBP2-13358-25ul |
25 μL |
€ 415.00 € 391.65 / 25µL Save € 23.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18477721
|
Novus Biologicals
NBP2-13358 |
0.1 mL |
€ 624.00 € 590.10 / 0.10mL Save € 33.90 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Syntaphilin Polyclonal antibody specifically detects Syntaphilin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Syntaphilin | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.2) and 40% Glycerol | |
| 9751 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| bA314N13.5, KIAA0374, MGC46096, syntaphilin | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: FPASNTYEKLLCGMEAGVQASCMQERAIQTDFVQYQPDLDTILEKVTQAQVCGTDPESGDRCPELDAHPSGPRDPNSAVVV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title