missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 18 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68220-1-IG-20UL
This item is not returnable.
View return policy
Description
The Syntaxin 18 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Syntaxin 18. This antibody recognizes human, mouse, rat, pig antigen. The Syntaxin 18 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| Syntaxin 18 | |
| Monoclonal | |
| 1000 μg/mL | |
| DKFZp686O15149; Ufe1 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| 39 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 2A2C9 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 53407 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction