missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Syntaxin 3 Recombinant Rabbit Monoclonal Antibody (K01_3K21)
GREENER_CHOICE

Product Code. p-200114115
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825553 100 μL
30828118 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825553 Supplier Invitrogen™ Supplier No. MA564338

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Syntaxin 3 Recombinant Monoclonal Antibody for Western Blot

The gene is a member of the syntaxin family. The encoded protein is targeted to the apical membrane of epithelial cells where it forms clusters and is important in establishing and maintaining polarity necessary for protein trafficking involving vesicle fusion and exocytosis. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaxin 3
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_3K21
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene Stx3
Gene Accession No. Q08849, Q13277, Q64704
Gene Alias STX3; STX3A; Syn-3; syntaxin 3; syntaxin 3A; syntaxin-3
Gene Symbols Stx3
Host Species Rabbit
Immunogen Peptide.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20908, 6809, 81802
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.