missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 4 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32227AF532
This item is not returnable.
View return policy
Description
Syntaxin 4 Polyclonal antibody specifically detects Syntaxin 4 in Human samples. It is validated for Western Blot
Specifications
| Syntaxin 4 | |
| Polyclonal | |
| Western Blot | |
| Renal carcinoma antigen NY-REN-31, STX4Ap35-2, syntaxin 4, syntaxin 4A (placental), syntaxin-4 | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Syntaxin 4 using the numbering given in entry NP_004595.2 (GeneID 6810). | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 6810 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction