missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94678-0.1ml
This item is not returnable.
View return policy
Description
Syntaxin 5 Polyclonal antibody specifically detects Syntaxin 5 in Rat samples. It is validated for Western Blot
Specifications
| Syntaxin 5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| STX5ASED5, syntaxin 5, syntaxin 5A, syntaxin-5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 55-355 of human STX5 (NP_003155.2). MSCRDRTQEFLSACKSLQTRQNGIQTNKPALRAVRQRSEFTLMAKRIGKDLSNTFAKLEKLTILAKRKSLFDDKAVEIEELTYIIKQDINSLNKQIAQLQDFVRAKGSQSGRHLQTHSNTIVVSLQSKLASMSNDFKSVLEVRTENLKQQRSRREQFSRAPVSALPLAPNHLGGGAVVLGAESHASKDVAIDMMDSRTSQQLQLIDEQDSYIQSRADTMQNIESTIVELGSIFQQLAHMVKEQEETIQRIDENVLGAQLDVEAAHSEILKYFQSVTSNRWLMVKIFLILIVFFIIFVVFLA | |
| 0.1 mL | |
| Signal Transduction | |
| 6811 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction