missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 8 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-00260CL1
This item is not returnable.
View return policy
Description
Syntaxin 8 Polyclonal antibody specifically detects Syntaxin 8 in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Syntaxin 8 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| CARB, CIP-1-associated regulator of cyclin B, syntaxin 8, syntaxin-8 | |
| Produced in rabbits immunized with purified, recombinant Human Syntaxin 8 (Uniprot#: Q9UNK0; Met1-Gly215) | |
| 0.1 mL | |
| Endosome Markers | |
| 9482 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction