missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GST Epitope Tag Antibody (1E10), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50415AF350
This item is not returnable.
View return policy
Description
GST Epitope Tag Monoclonal antibody specifically detects GST Epitope Tag in Epitope Tag samples. It is validated for Western Blot
Specifications
| GST Epitope Tag | |
| Monoclonal | |
| Alexa Fluor 350 | |
| EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase | |
| GST protein | |
| 0.1 mL | |
| Asthma, Cancer, Epitope Tags, Immunology | |
| 2944 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| 1E10 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Tag | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction