missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin-BP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-3451-0.025mg
This item is not returnable.
View return policy
Description
Syntaxin-BP1 Polyclonal specifically detects Syntaxin-BP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| Syntaxin-BP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| STXBP1 | |
| Synthetic Peptide: V(58) E D I N K R R E P I P S(70) | |
| 0.025 mg | |
| Primary | |
| Munc 18 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
| EIEE4, FLJ37475, hUNC18, MUNC18-1, N-Sec1, NSEC1, P67, Protein unc-18 homolog 1, Protein unc-18 homolog A, RBSEC1, syntaxin binding protein 1, syntaxin-binding protein 1, Unc18-1, unc-18A, UNC18A, UNC18neuronal SEC1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6812 | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction