missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin-BP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20526
This item is not returnable.
View return policy
Description
Syntaxin-BP1 Polyclonal specifically detects Syntaxin-BP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Syntaxin-BP1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| STXBP1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of humanSyntaxin-BP1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 6812 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.96mg/mL | |
| Western Blot 1:500-1:20000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| EIEE4, FLJ37475, hUNC18, MUNC18-1, N-Sec1, NSEC1, P67, Protein unc-18 homolog 1, Protein unc-18 homolog A, RBSEC1, syntaxin binding protein 1, syntaxin-binding protein 1, Unc18-1, unc-18A, UNC18A, UNC18neuronal SEC1 | |
| Rabbit | |
| 68 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse. Species reactivity based on sequence identity: Rat (100%), Rhesus Monkey (100%), Bovine (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction