Learn More
Invitrogen™ Syntenin 1 Monoclonal Antibody (GT1523)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA531572
Product Code. 16385004
  • € 522.00 / 100µL
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Pig (100%), Chicken (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Positive Control: U87-MG, rat hippocampus. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene was initially identified as a molecule linking syndecan-mediated signaling to the cytoskeleton. The syntenin protein contains tandemly repeated PDZ domains that bind the cytoplasmic, C-terminal domains of a variety of transmembrane proteins. This protein may also affect cytoskeletal-membrane organization, cell adhesion, protein trafficking, and the activation of transcription factors. The protein is primarily localized to membrane-associated adherens junctions and focal adhesions but is also found at the endoplasmic reticulum and nucleus. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Syntenin 1
Monoclonal
1 mg/mL
PBS with no preservative
O00560, Q9JI92
SDCBP
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Syntenin 1. The exact sequence is proprietary.
100 μL
Primary
Human, Rat
Antibody
IgG2a
Western Blot, Immunocytochemistry
GT1523
Unconjugated
SDCBP
MDA9; MDA-9; melanoma differentiation associated protein-9; melanoma differentiation-associated protein 9; Pro-TGF-alpha cytoplasmic domain-interacting protein 18; sb:eu862; Scaffold protein Pbp1; sdcbp; si:dkey-235k4.1; ST1; Sycl; syndecan binding protein; syndecan binding protein (syntenin); syndecan interacting protein; syndecan-binding protein; syndecan-binding protein 1; syntenin; syntenin 1; syntenin-1; syntenin-b; TACIP18; Unknown (protein for MGC:128349)
Mouse
Protein A
RUO
6386, 83841
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.