Learn More
Syntenin-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22399-1-AP-150UL
 View more versions of this product
Product Code. 16801925
  • € 410.00 / 150µL
Description
The protein encoded by this gene was initially identified as a molecule linking syndecan-mediated signaling to the cytoskeleton. The syntenin protein contains tandemly repeated PDZ domains that bind the cytoplasmic, C-terminal domains of a variety of transmembrane proteins. This protein may also affect cytoskeletal-membrane organization, cell adhesion, protein trafficking, and the activation of transcription factors.
Specifications
Specifications
Syntenin-1
Polyclonal
Unconjugated
SDCBP
MDA 9, MDA9, Scaffold protein Pbp1, SDCBP, ST1, SYCL, Syndecan binding protein 1, Syntenin-1, TACIP18
Rabbit
Antigen Affinity Chromatography
RUO
6386
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00560
SDCBP
Syntenin-1 Fusion Protein Ag18078
150 μL