missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Syntenin 1 Recombinant Rabbit Monoclonal Antibody (23GB2185), Invitrogen™
GREENER_CHOICE

Product Code. p-200087863
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30087585 100 μL 100µL
30089547 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30087585 Supplier Invitrogen™ Supplier No. MA553244

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

The protein encoded by this gene was initially identified as a molecule linking syndecan-mediated signaling to the cytoskeleton. The syntenin protein contains tandemly repeated PDZ domains that bind the cytoplasmic, C-terminal domains of a variety of transmembrane proteins. This protein may also affect cytoskeletal-membrane organization, cell adhesion, protein trafficking, and the activation of transcription factors. The protein is primarily localized to membrane-associated adherens junctions and focal adhesions but is also found at the endoplasmic reticulum and nucleus. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntenin 1
Applications Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2185
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SDCBP
Gene Accession No. O00560
Gene Alias MDA9; MDA-9; melanoma differentiation associated protein-9; melanoma differentiation-associated protein 9; Pro-TGF-alpha cytoplasmic domain-interacting protein 18; sb:eu862; Scaffold protein Pbp1; sdcbp; si:dkey-235k4.1; ST1; Sycl; syndecan binding protein; syndecan binding protein (syntenin); syndecan interacting protein; syndecan-binding protein; syndecan-binding protein 1; syntenin; syntenin 1; syntenin-1; syntenin-b; TACIP18; Unknown (protein for MGC:128349)
Gene Symbols SDCBP
Host Species Rabbit
Immunogen A synthesized peptide derived from human Syntenin (2-79AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6386
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.