missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Syntenin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 180.00 - € 476.00
Especificaciones
| Antigen | Syntenin 2 |
|---|---|
| Concentration | 0.64 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
17750854
![]() |
Invitrogen™
PA5119451 |
200 μL |
€ 476.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17722964
|
Invitrogen™
PA5121564 |
20 μL |
€ 180.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
Syntenin 2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
SDCBP2 contains 2 PDZ (DHR) domains. The function of SDCBP2 remains unknown.Especificaciones
| Syntenin 2 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| SDCBP2 | |
| SDCBP2; Similar to TACIP18; SITAC; SITAC18; ST-2; syndecan binding protein (syntenin) 2; syndecan binding protein 2; syndecan-binding protein 2; Syntenin-2 | |
| SDCBP2 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.64 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q99JZ0, Q9H190 | |
| 228765, 27111 | |
| Recombinant protein Syntenin 2. The antigen corresponds to amino acid range 1-292 of the target protein. | |
| Antibody |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto