missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntrophin Mouse anti-Amphibian, Canine, Chicken, Human, Mouse, Rat, Clone: 1351, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB600-1294-0.025mg
This item is not returnable.
View return policy
Description
Syntrophin Monoclonal specifically detects Syntrophin in Human, Mouse, Rat, Amphibian, Canine, Chicken samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Syntrophin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.1 - 1 μg/mL, Flow Cytometry 0.5 μg/ 106 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 2 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
| Q13425 | |
| SNTA1 | |
| Whole purified syntrophin from Torpedo californica electric organ postsynaptic membrane. | |
| 0.025 mg | |
| Primary | |
| This antibody detects the alpha 1, beta 1 and beta 2 subunits of Syntrophin. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 1351 | |
| Unconjugated | |
| PBS,1mg/ml BSA with 0.05% Sodium Azide | |
| 59 kDa dystrophin-associated protein A1 acidic component 1, dJ1187J4.5, LQT12, Pro-TGF-alpha cytoplasmic domain-interacting protein 1, SNT1alpha-1-syntrophin, syntrophin, alpha 1 (dystrophin-associated protein A1, 59kD, acidic component), syntrophin, alpha 1 (dystrophin-associated protein A1, 59kDa, acidic component), syntrophin-1, TACIP1acidic alpha 1 syntrophin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6640 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction