missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Synuclein alpha/beta Monoclonal Antibody (Syn 202)

Product Code. 10622764
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10622764 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10622764 Supplier Invitrogen™ Supplier No. 328200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Synuclein alpha/beta Monoclonal Antibody for Western Blot, ICC/IF, IHC, IP, ELISA

Synuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer's (AD), Parkinson's PD) and breast cancer. In AD, a peptide derived from alpha synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of alpha synuclein has been linked to many familial cases of PD. In breast cancer increased expression of gamma synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synuclein alpha/beta
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone Syn 202
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Snca
Gene Accession No. O55042, P37377, P37840, Q16143, Q63754, Q91ZZ3
Gene Alias AI838531; alphaSYN; alpha-synuclein; betaSYN; Beta-synuclein; NACP; non A-beta component of AD amyloid; non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; Phosphoneuroprotein 14; PNP 14; SNCA; Sncb; Syn; synuclein alpha; synuclein alpha-140; synuclein beta; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor); synuclein, beta
Gene Symbols Snca, SNCB
Host Species Mouse
Immunogen Recombinant human alpha- and beta-synuclein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104069, 113893, 20617, 29219, 6620, 6622
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.