missing translation for 'onlineSavingsMsg'
Learn More

SYP Monoclonal Antibody (OTI1G3), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA506068S

Encompass_Preferred

 View more versions of this product

Product Code. 17555352

  • € 242.00 / 30µL
Only null left
Estimated Shipment: 15-01-2026
Add to basket
This item is not returnable. View return policy

Description

Description

SYP Monoclonal antibody specifically detects SYP in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

SYP
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P08247
Mouse
Affinity chromatography
RUO
6855
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
OTI1G3
Unconjugated
Syp
A230093K24Rik; AI848995; BM89 antigen; I79_012252; Major synaptic vesicle protein p38; MRX96; MRXSYP; p38; Syn; Synaptophysin; Syp; Syp I; Syp1
Full length human recombinant protein of human SYP produced in HEK293T cell.
30 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.