missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYP Rabbit anti-Human, Clone: RM258, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB15137.100uL
This item is not returnable.
View return policy
Description
Rabbit monoclonal antibody raised against synthetic peptide of human SYP.
- High specificity
- High reactivity
Specifications
| SYP | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:500) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
| P08247 | |
| Rabbit | |
| Protein A affinity purification | |
| RUO | |
| 6855 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| RM258 | |
| Rabbit monoclonal antibody raised against synthetic peptide of human SYP. | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| SYP | |
| A synthetic peptide corresponding to C-terminus of human SYP. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction