missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYPL1 Recombinant Rabbit Monoclonal Antibody (23GB4675), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 217.00 - € 590.00
Specifications
| Antigen | SYPL1 |
|---|---|
| Clone | 23GB4675 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089253
![]() |
Invitrogen™
MA553786 |
100 μL |
€ 590.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089591
|
Invitrogen™
MA553787 |
20 μL |
€ 217.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SYPL1 Recombinant Monoclonal antibody specifically detects SYPL1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| SYPL1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q16563 | |
| 6856 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4675 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI314763; AI604763; D12Ertd446e; H-SP1; Pan I; PanI; Pantophysin; Pphn; synaptophysin like 1; synaptophysin-like 1; synaptophysin-like protein; synaptophysin-like protein 1; SYPL; Sypl1 | |
| A synthesized peptide derived from human SYPL1 (150-259AA). | |
| Primary | |
| -20°C | |
| SYPL1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title