Learn More
Hamilton™ Custom Cemented Needle Syringes
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
25 mL
Product Type:
Gastight
Syringe Needle
Unit Size:
Each
Description
- Ideal for dispensing volumes from 1 up to 500 μL
- A variety of volumes, lengths, and gauges available
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Needle Gauge | Custom |
| Diameter (Metric) Inner | 0.46 mm |
| Diameter (Metric) Outer | 6.60 mm |
| Product Type | Gastight |
| Max. Pressure | 68.9 bar |
| Max. Temperature (Metric) | 50°C |
| Model | 1701 |
| Min. Temperature (Metric) | 10°C |
| Quantity | 1 Ea. |
| Show More |