Learn More
Poulten & Graf GmbH FORTUNA™ Gas Syringe
Click to view available options
Volume (Metric):
50 mL
100 mL
Unit Size:
Each
Description
Made of borosilicate glass
Amber graduations
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Diameter (Metric) Inner Needle | 2.5 mm |
| Diameter (Metric) Outer Needle | 7.5 mm |
| Product Type | Syringe |
| Quantity | 1/Pk. |
| Volume (Metric) | 100 mL |