missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYT11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16579801
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16579801 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16579801 Supplier Invitrogen™ Supplier No. PA596970

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: WSCCHQQAEK KQKNPPYKFI HMLKGISIYP ETLSNKKKII KVRRDKDGPG REGGRRNLLV DAAEAGLLSR DKDPRGPSSG SCIDQLPIKM DYGEELRSPI TSLTPGESKT TSPSSPEEDV MLGSLTFSVD YNFPKKALVV TIQEAHGLPV MDDQTQGSDP YIKM; Positive Samples: U-251MG, Mouse brain, Rat brain; Cellular Location: Cell junction, Cytoplasmic vesicle, Membrane, Single-pass membrane protein, secretory vesicle, synapse, synaptic vesicle membrane.

This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that are known calcium sensors and mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. The encoded protein is also a substrate for ubiquitin-E3-ligase parkin. The gene has previously been referred to as synaptotagmin XII but has been renamed synaptotagmin XI to be consistent with mouse and rat official nomenclature. [provided by RefSeq, Apr 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYT11
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SYT11
Gene Accession No. O08835, Q9BT88, Q9R0N3
Gene Alias 1500004A13Rik; 3632445O20Rik; 5430404N14Rik; 6530420C11Rik; AI851970; KIAA0080; synaptotagmin 11; synaptotagmin 12; synaptotagmin XI; Synaptotagmin-11; SYT11; SYT12; SytXI
Gene Symbols SYT11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 37-200 of human SYT11 (NP_6894933).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229521, 23208, 60568
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.