missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYT13 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13496667
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13496667 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13496667 Supplier Invitrogen™ Supplier No. PA532101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, U87-MG, SK-N-SH. Predicted reactivity: Mouse (89%), Rat (88%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SYT13 belongs to the large synaptotagmin protein family. All synaptotagmins show type I membrane topology, with an extracellular N terminus, a single transmembrane region, and a cytoplasmic C terminus containing tandem C2 domains. Major functions of synaptotagmins include vesicular traffic, exocytosis, and secretion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYT13
Applications Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SYT13
Gene Accession No. Q7L8C5
Gene Alias 5730409J20Rik; AI549909; Kiaa1427; mKIAA1427; synaptotagmin 13; synaptotagmin XIII; Synaptotagmin-13; SYT13; SytXIII
Gene Symbols SYT13
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SYT13. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57586
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.