missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYT16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101919
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SYT16.
SYT16 is a protein coding gene. May be involved in the trafficking and exocytosis of secretory vesicles in non-neuronal tissues. Is Ca(2+)-independent.
Specifications
| SYT16 | |
| Polyclonal | |
| Unconjugated | |
| SYT16 | |
| chr14 synaptotagmin; CHR14SYT; CTD-2277K2.1; STREP14; Synaptotagmin 14-like protein; synaptotagmin 16; synaptotagmin XIV-related protein; synaptotagmin XVI; synaptotagmin-16; SYT14L; syt14r; SYT16; yt14r | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 83851 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q17RD7 | |
| SYT16 | |
| A synthesized peptide derived from human SYT16(Accession Q17RD7), corresponding to amino acid residues L232-H282. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction