Learn More
SYT4 Monoclonal Antibody (OTI3E10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA505846S
Encompass_Preferred
 View more versions of this product
Product Code. 17564862
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
SYT4 Monoclonal antibody specifically detects SYT4 in Canine, Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot
Specifications
Specifications
SYT4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O09110, P46734
Mouse
Affinity chromatography
RUO
26397, 489547, 5606
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunocytochemistry, Western Blot
OTI3E10
Unconjugated
MAP2K3
HsT1192; KIAA1342; synaptotagmin 4; synaptotagmin IV; Synaptotagmin4; Synaptotagmin-4; Syt3; Syt4; SytIV
Full length human recombinant protein of human MAP2K3 produced in HEK293T cell.
30 μL
Primary
Canine, Human, Mouse