missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYTL4 Monoclonal Antibody (OTI1H8), TrueMAB™, OriGene
Description
This gene encodes a member of the synaptotagmin like protein family. Members of this family are characterized by an N-terminal Rab27 binding domain and C-terminal tandem C2 domains. The encoded protein binds specific small Rab GTPases and is involved in intracellular membrane trafficking. This protein binds Rab27 and may be involved in inhibiting dense core vesicle exocytosis. Alternate splicing results in multiple transcript variants that encode the same protein.
Specifications
Specifications
| Antigen | SYTL4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1H8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | SYTL4 |
| Gene Accession No. | Q96C24 |
| Gene Alias | SLP4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?