missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYTL4 Monoclonal Antibody (OTI3C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810588
This item is not returnable.
View return policy
Description
This gene encodes a member of the synaptotagmin like protein family. Members of this family are characterized by an N-terminal Rab27 binding domain and C-terminal tandem C2 domains. The encoded protein binds specific small Rab GTPases and is involved in intracellular membrane trafficking. This protein binds Rab27 and may be involved in inhibiting dense core vesicle exocytosis. Alternate splicing results in multiple transcript variants that encode the same protein.Specifications
| SYTL4 | |
| Monoclonal | |
| Unconjugated | |
| SYTL4 | |
| SLP4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 94121 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3C8 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96C24 | |
| SYTL4 | |
| Full length human recombit protein of human SYTL4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction