Learn More
SYTL4 Monoclonal Antibody (OTI3C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810588S
This item is not returnable.
View return policy
Description
SYTL4 Monoclonal antibody specifically detects SYTL4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| SYTL4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6NUP7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57718 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3C8 | |
| Unconjugated | |
| PPP4R4 | |
| Exophilin-2; Gph; Granuphilin; granuphilin b; granuphilin-a; granuphilin-b; Slp4; synaptotagmin like 4; synaptotagmin-like 4; synaptotagmin-like 4 (granuphilin-a); synaptotagmin-like protein 4; Sytl4 | |
| Full length human recombinant protein of human PPP4R4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |