missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYTL4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539490
This item is not returnable.
View return policy
Description
This gene encodes a member of the synaptotagmin like protein family. Members of this family are characterized by an N-terminal Rab27 binding domain and C-terminal tandem C2 domains. The encoded protein binds specific small Rab GTPases and is involved in intracellular membrane trafficking. This protein binds Rab27 and may be involved in inhibiting dense core vesicle exocytosis. Alternate splicing results in multiple transcript variants that encode the same protein. [provided by RefSeq].
Specifications
| SYTL4 | |
| Polyclonal | |
| Unconjugated | |
| SYTL4 | |
| SLP4, synaptotagmin-like 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140594, 27359, 94121 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96C24, Q9R0Q1, Q8VHQ7 | |
| SYTL4 | |
| A synthetic peptide derived from the internal region of human SYTL4 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction