missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYTL5 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81817AF647
This item is not returnable.
View return policy
Description
SYTL5 Polyclonal antibody specifically detects SYTL5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SYTL5 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 647 | |
| exophilin 9, synaptotagmin-like 5 | |
| SYTL5 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human SYTL5. The immunogen is located within amino acids 30 - 80 of SYTL5. Amino Acid Squence: LEAKRRSGKTQQEASRV | |
| 0.1 mL | |
| Signal Transduction | |
| 94122 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction