missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYTOX AADvanced™ Ready Flow™ Reagent

Product Code. 15952344
Change view
Click to view available options
Quantity:
120 Reactions
Unit Size:
120 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15952344 120 Reactions 120 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15952344 Supplier Invitrogen™ Supplier No. R37173

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SYTOX™ AADvanced Ready Flow Reagent is a high-affinity nucleic acid stain for the detection of dead cells in flow cytometry.

  • A high-affinity nucleic acid stain for the detection of dead cells in flow cytometry
  • SYTOX AADvanced Ready Flow Reagent is a ready-to-use, direct-to-sample dead cell stain that emits red fluorescence when bound to DNA
  • It is spectrally similar to 7-AAD, but with rapid uptake kinetics and relatively low CVs
  • It penetrates cells more efficiently than 7-AAD, providing better separation of live and dead cells
  • Add 2 drops form the dropper bottle directly into 1 x 106 cells, incubate, and analyze
  • The dye has excitation/emission maxima of 546/647 nm
  • No need to dilute, weigh, or pipette
  • Stable at room temperature
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Impermeant
Color Red
Content And Storage Includes 2 dropper bottles, store at room temperature.
Description SYTOX AADvanced™ Ready Flow™ Reagent
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Reagent
Dye Type Cell-Impermeant
Emission 647
Excitation Wavelength Range 546 nm
Form Solution
Format Bottle(s)
Product Line AADvanced, Ready Flow
Quantity 120 Reactions
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Sub Cellular Localization Nucleus
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.