missing translation for 'onlineSavingsMsg'
Learn More

T-bet/TBX21 Antibody (525803), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18592237 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18592237 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18592237 Supplier Novus Biologicals Supplier No. FAB5385X

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen T-bet/TBX21
Applications Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry
Classification Monoclonal
Clone 525803
Conjugate Alexa Fluor 532
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM
Host Species Mouse
Immunogen E. coli-derived recombinant human T-bet, Glu326-Asn535, Accession # Q9UL17
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 30009
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.