missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T-bet/TBX21 Antibody (525831), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB53851MFV500
This item is not returnable.
View return policy
Description
T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| T-bet/TBX21 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Immunology | |
| 30009 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 525831 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM | |
| E. coli-derived recombinant human T-bet/TBX21, Glu326-Asn535, Accession # Q9UL17 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction