missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ T-Cell Receptor Signaling Phospho-Specific Array

Product Code. 16052445
Change view
Click to view available options
Quantity:
1 Kit (2 Samples)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16052445 1 Kit (2 Samples) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16052445 Supplier Abnova™ Supplier No. AA0040.1Kit(2Samples)

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

T-Cell Receptor Signaling Phospho-Specific Array includes 188 highly specific and well-characterized phosphorylation antibodies in the T-cell receptor signaling pathway.

  • This product must be accompanied with Antibody Array Detection Kit
  • The arrays are printed on standard-size coated glass microscope slides and can be scanned on all microarray scanners that are compatible with 76 x 25 x 1 mm slides
  • Suitable Sample: Cultured Cells, Frozen Tissue, FFPE Tissue, Serum

Specifications

Content And Storage 4°C
Detection Method Fluorometric
Quantity 1 Kit (2 Samples)
For Use With (Application) Profiling
Product Type Array/Library
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.