missing translation for 'onlineSavingsMsg'
Learn More
Learn More
t-Plasminogen Activator/tPA Antibody (OTI3H3), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74568UV
This item is not returnable.
View return policy
Description
t-Plasminogen Activator/tPA Monoclonal specifically detects t-Plasminogen Activator/tPA in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| t-Plasminogen Activator/tPA | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PLAT | |
| Recombinant protein fragment corresponding to amino acids 157-426 of human t-Plasminogen Activator/tPA (NP_000921) produced in E.coli. | |
| 0.1 mL | |
| 5327.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3H3 | |
| Western Blot, Immunohistochemistry | |
| alteplase, DKFZp686I03148, EC 3.4.21, EC 3.4.21.68, plasminogen activator, tissue, plasminogen activator, tissue type, reteplase, tissue plasminogen activator (t-PA), tissue-type plasminogen activator, t-PA, TPA, t-plasminogen activator | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction