missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ T4S20 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA566779

Product Code. 15939464

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: IHPESFNLQWF FNDSCAPPTG FNKPTSNDTM ASGWRASSFH FDSEENKHRL IHFSV Highest antigen sequence identity to the following orthologs - mouse 52%, rat 52%.

Polytopic transmembrane protein that inhibits regulated intramembrane proteolysis (RIP) of CREB3L1, inhibiting its activation and the induction of collagen synthesis (PubMed:25310401, PubMed:27499293). In response to ceramide, which alters TM4SF20 membrane topology, stimulates RIP activation of CREB3L1 (PubMed:27499293). Ceramide reverses the direction through which transmembrane helices are translocated into the endoplasmic reticulum membrane during translation of TM4SF20, this mechanism is called 'regulated alternative translocation' (RAT) and regulates the function of the transmembrane protein (PubMed:27499293).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

T4S20
Polyclonal
Unconjugated
TM4SF20
PRO994; SLI5; TCCE518; TM4SF20; transmembrane 4 L six family member 20; transmembrane 4 L6 family member 20; UNQ518/PRO994
Rabbit
Antigen affinity chromatography
RUO
79853
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q53R12
TM4SF20
Recombinant protein fragment of Human TM4SF20 (Product #RP-104287)
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.