missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ T4S20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562495
This item is not returnable.
View return policy
Description
Immunogen sequence: IHPESFNLQW FFNDSCAPPT GFNKPTSNDT MASGWRASSF HFDSEENKHR LIHFSV Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 52%.
Polytopic transmembrane protein that inhibits regulated intramembrane proteolysis (RIP) of CREB3L1, inhibiting its activation and the induction of collagen synthesis (PubMed:25310401, PubMed:27499293). In response to ceramide, which alters TM4SF20 membrane topology, stimulates RIP activation of CREB3L1 (PubMed:27499293). Ceramide reverses the direction through which transmembrane helices are translocated into the endoplasmic reticulum membrane during translation of TM4SF20, this mechanism is called 'regulated alternative translocation' (RAT) and regulates the function of the transmembrane protein (PubMed:27499293).
Specifications
| T4S20 | |
| Polyclonal | |
| Unconjugated | |
| TM4SF20 | |
| PRO994; SLI5; TCCE518; TM4SF20; transmembrane 4 L six family member 20; transmembrane 4 L6 family member 20; UNQ518/PRO994 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79853 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53R12 | |
| TM4SF20 | |
| Recombinant protein corresponding to Human T4S20. Recombinant protein control fragment (Product #RP-104287). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction