missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAB2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | TAB2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230765
|
Novus Biologicals
NBP3-38625-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227709
|
Novus Biologicals
NBP3-38625-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAB2 Polyclonal antibody specifically detects TAB2 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| TAB2 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 23118 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ21885, KIAA0733CHTD2, MAP3K7IP2mitogen-activated protein kinase kinase kinase 7 interacting protein 2, Mitogen-activated protein kinase kinase kinase 7-interacting protein 2, TAB-2, TAK1-binding protein 2, TGF-beta activated kinase 1/MAP3K7 binding protein 2, TGF-beta-activated kinase 1 and MAP3K7-binding protein 2, TGF-beta-activated kinase 1-binding protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 510-620 of human TAB2 (NP_055908.1).,, Sequence:, TLAHVDRISETRKLSMGSDDAAYTQALLVHQKARMERLQRELEIQKKKLDKLKSEVNEMENNLTRRRLKRSNSISQIPSLEEMQQLRSCNRQLQIDIDCLTKEIDLFQARG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title