missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAB2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17189583
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17189583 100 μL 100µL
17159573 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17189583 Supplier Invitrogen™ Supplier No. PA5111670

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: HSGWVSQFNP MNPQQVYQPS QPGPWTTCPA SNPLSHTSSQ QPNQQGHQTS HVYMPISSPT TSQPPTIHSS Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 93%.

TAB2 is an activator of MAP3K7/TAK1, which is required for the IL-1 induced activation NF-kappaB and MAPK8/JNK. This protein forms a kinase complex with TRAF6, MAP3K7 and TAB1, thus serves as an adaptor linking MAP3K7 and TRAF6. This protein, TAB1, and MAP3K7 also participate in the signal transduction induced by TNFSF11/RANKL through the activation of the receptor activator of NF-kappaB (TNFRSF11A/RANK), which may regulate the development and function of osteoclasts. Recent experiments have shown that TAB2 and the related protein TAB3 constitutively interact with the autophagy mediator Beclin-1; upon induction of autophagy, these proteins dissociate from Beclin-1 and bind TAK1. Overexpression of TAB2 and TAB3 inhibit autophagy, while their depletion triggers it, suggesting that TAB2 and TAB3 act as a control point for autophagy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAB2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tab2
Gene Accession No. Q9NYJ8
Gene Alias 1110030N06Rik; A530078N03Rik; CHTD2; FLJ21885; KIAA0733; MAP3K7IP2; mitogen activated protein kinase kinase kinase 7 interacting protein 2; mitogen-activated protein kinase kinase kinase 7 interacting protein 2; mitogen-activated protein kinase kinase kinase 7-interacting protein 2; mKIAA0733; OTTHUMP00000017388; Tab2; TAB-2; Tak1 binding protein 2; TAK1-binding protein 2; TGF-beta activated kinase 1 (MAP3K7) binding protein 2; TGF-beta activated kinase 1/MAP3K7 binding protein 2; TGF-beta-activated kinase 1 and MAP3K7-binding protein 2; TGF-beta-activated kinase 1-binding protein 2
Gene Symbols Tab2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TAB2. Recombinant protein control fragment (Product #RP-107803).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23118
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.