missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAC1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5110792

Product Code. 17168713

  • € 630.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: WSDWYDSDQI KEELPEPFEH LLQRIARRPK PQQFFGLMGK RDADSSIEKQ VALLKALYGH GQISHKRHKT DSFVGLMGKR ALNSVAYERS AMQNYERRR Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 96%.

This gene encodes four products of the tachykinin peptide hormone family, substance P and neurokinin A, as well as the related peptides, neuropeptide K and neuropeptide gamma. These hormones are thought to function as neurotransmitters which interact with nerve receptors and smooth muscle cells. They are known to induce behavioral responses and function as vasodilators and secretagogues. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TAC1
Polyclonal
Unconjugated
TAC1
4930528L02Rik; C-terminal-flanking peptide; Hs.2563; neurokinin 1; neurokinin 2; neurokinin A; neurokinin alpha; neuromedin L; Neuropeptide gamma; neuropeptide K; NK1; NK-1; NK2; NKA; NKNA; NPK; PPT; Ppt5fl; PPTA; PPT-A; PPTA3; preprotachykinin; preprotachykinin A; protachykinin; protachykinin-1; RATPPTA3; SP; substance K; Substance P; TAC; Tac1; Tac2; Tachykinin (substance P, neurokinin A, neuropeptide K, neuropeptide gamma); tachykinin 1; tachykinin 2; tachykinin precursor 1; tachykinin, precursor 1; tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma)
Rabbit
Antigen affinity chromatography
RUO
6863
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P20366
TAC1
Recombinant Protein corresonding to Human TAC1. Recombinant protein control fragment (Product #RP-90655).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.