missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACC1 Antibody (OTI1H2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74429F
This item is not returnable.
View return policy
Description
TACC1 Monoclonal specifically detects TACC1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| TACC1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| TACC1 | |
| Full length recombinant protein of human TACC1 (NP_001116296) produced in HEK293T cells. | |
| 0.1 mL | |
| 6867.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI1H2 | |
| Immunohistochemistry | |
| DKFZp686K18126, FLJ42304, Ga55, Gastric cancer antigen Ga55, KIAA1103, taxin 1, taxin-1, transforming acidic coiled-coil-containing protein 1, transforming, acidic coiled-coil containing protein 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction