missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACC2 Antibody (OTI3C4), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74430IR
This item is not returnable.
View return policy
Description
TACC2 Monoclonal specifically detects TACC2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TACC2 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TACC2 | |
| Recombinant protein fragment corresponding to amino acids 727-1026 of human TACC2 (NP_008928) produced in E.coli. | |
| 0.1 mL | |
| 10579.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3C4 | |
| Western Blot, Immunohistochemistry | |
| anti zuai-1, AZU-1ECTACC, EC 2.4.2.10, EC 4.3.2.1, EC 6.3.5.2, transforming acidic coiled-coil-containing protein 2, transforming, acidic coiled-coil containing protein 2, Zuai-1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction