missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TACC2 Monoclonal Antibody (OTI1C3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526619
This item is not returnable.
View return policy
Description
TACC2 Monoclonal Antibody for Western Blot, IHC (P)
Transforming acidic coiled-coil proteins are a conserved family of centrosome- and microtubule-interacting proteins that are implicated in cancer. This gene encodes a protein that concentrates at centrosomes throughout the cell cycle. This gene lies within a chromosomal region associated with tumorigenesis. Expression of this gene is induced by erythropoietin and is thought to affect the progression of breast tumors. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| TACC2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95359 | |
| TACC2 | |
| Human recombinant protein fragment corresponding to amino acids 727-1026 of TACC2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C3 | |
| Unconjugated | |
| TACC2 | |
| anti zuai-1; Anti-Zuai-1; AZU-1; ECTACC; mKIAA4180; MNCb-3527; RP11-296H2.1; TACC2; transforming acidic coiled coil 2; transforming acidic coiled-coil containing protein 2; transforming acidic coiled-coil containing protein 2 variant 3; transforming acidic coiled-coil-containing protein 2; transforming, acidic coiled-coil containing protein 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10579 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction