Learn More
TACC2 Monoclonal Antibody (OTI2G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA804148S
Encompass_Preferred
 View more versions of this product
Product Code. 17583533
  • € 242.00 / 30µL
Description
TACC2 Monoclonal antibody specifically detects TACC2 in Human samples. It is validated for Western Blot
Specifications
Specifications
TACC2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O95359
Mouse
Affinity chromatography
RUO
10579
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI2G3
Unconjugated
TACC2
anti zuai-1; Anti-Zuai-1; AZU-1; ECTACC; mKIAA4180; MNCb-3527; RP11-296H2.1; TACC2; transforming acidic coiled coil 2; transforming acidic coiled-coil containing protein 2; transforming acidic coiled-coil containing protein 2 variant 3; transforming acidic coiled-coil-containing protein 2; transforming, acidic coiled-coil containing protein 2
Human recombinant protein fragment corresponding to amino acids 727-1026 of human TACC2 produced in E.coli.