missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACC2 Monoclonal Antibody (OTI4D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804146S
This item is not returnable.
View return policy
Description
TACC2 Monoclonal antibody specifically detects TACC2 in Human samples. It is validated for Western BlotSpecifications
| TACC2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95359 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10579 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4D10 | |
| Unconjugated | |
| TACC2 | |
| anti zuai-1; Anti-Zuai-1; AZU-1; ECTACC; mKIAA4180; MNCb-3527; RP11-296H2.1; TACC2; transforming acidic coiled coil 2; transforming acidic coiled-coil containing protein 2; transforming acidic coiled-coil containing protein 2 variant 3; transforming acidic coiled-coil-containing protein 2; transforming, acidic coiled-coil containing protein 2 | |
| Human recombinant protein fragment corresponding to amino acids 727-1026 of human TACC2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction