missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACC3 Antibody (OTI6F1), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74436MFV610
This item is not returnable.
View return policy
Description
TACC3 Monoclonal antibody specifically detects TACC3 in Human, Monkey, Primate samples. It is validated for Flow Cytometry
Specifications
| TACC3 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Monkey, Primate | |
| Purified |
| Flow Cytometry | |
| OTI6F1 | |
| Flow Cytometry | |
| ERIC1ERIC-1, MGC117382, MGC133242, transforming acidic coiled-coil-containing protein 3, transforming, acidic coiled-coil containing protein 3 | |
| Full length human recombinant protein of human TACC3(NP_006333) produced in HEK293T cell. | |
| 0.1 mL | |
| Apoptosis, Cancer, Mitotic Regulators, Tumor Suppressors | |
| 10460 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction