missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACC3 Antibody (OTI6F1), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74436PCP
This item is not returnable.
View return policy
Description
TACC3 Monoclonal specifically detects TACC3 in Human, Primate, Monkey samples. It is validated for Flow Cytometry.
Specifications
| TACC3 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| TACC3 | |
| Full length recombinant protein of human TACC3 (NP_006333) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Primate, Monkey | |
| IgG1 |
| Flow Cytometry | |
| OTI6F1 | |
| Flow Cytometry | |
| ERIC1ERIC-1, MGC117382, MGC133242, transforming acidic coiled-coil-containing protein 3, transforming, acidic coiled-coil containing protein 3 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer, Mitotic Regulators, Tumor Suppressors | |
| 10460.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction